Imprimer

Anti-HspB8 Mouse Monoclonal Antibody [clone: 3C12-H11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326394EA 645 EUR
ORIGTA326394
Anti-HspB8 Mouse Monoclonal Antibody [clone: 3C12-H11]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HspB8
Clonality: Monoclonal
Clone: 3C12-H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR