Imprimer

Anti-HIF1AN Mouse Monoclonal Antibody [clone: FIH 162c]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326397EA 554 EUR
ORIGTA326397
Anti-HIF1AN Mouse Monoclonal Antibody [clone: FIH 162c]
Recommended Dilutions: Western Blot: 10?g/ml

Type: Primary
Antigen: HIF1AN
Clonality: Monoclonal
Clone: FIH 162c
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR