Imprimer

Anti-AMIGO1 Mouse Monoclonal Antibody [clone: S86-36]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326398EA 645 EUR
ORIGTA326398
Anti-AMIGO1 Mouse Monoclonal Antibody [clone: S86-36]
Anticorps
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: AMIGO1
Clonality: Monoclonal
Clone: S86-36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR