Imprimer

Anti-KCNQ2 Mouse Monoclonal Antibody [clone: S26A-23]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326403EA 574 EUR
ORIGTA326403
Anti-KCNQ2 Mouse Monoclonal Antibody [clone: S26A-23]
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: KCNQ2
Clonality: Monoclonal
Clone: S26A-23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR