Imprimer

Anti-CDC37 Rabbit Polyclonal Antibody [clone: N/A]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326411EA 505 EUR
ORIGTA326411
Anti-CDC37 Rabbit Polyclonal Antibody [clone: N/A]
Anticorps
Type: Primary
Antigen: CDC37
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR