Imprimer

Anti-BCL2L11 Rabbit Polyclonal Antibody [clone: N/A]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326413EA 645 EUR
ORIGTA326413
Anti-BCL2L11 Rabbit Polyclonal Antibody [clone: N/A]
Anticorps
Recommended Dilutions: Western Blot: 1:400 Immunochemistry: 1:100

Type: Primary
Antigen: BCL2L11
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR