Imprimer

Anti-HSF1 Rat Monoclonal Antibody [clone: 10H8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326414EA 500 EUR
ORIGTA326414
Anti-HSF1 Rat Monoclonal Antibody [clone: 10H8]
Anticorps
Recommended Dilutions: 1?g/ml

Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone: 10H8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Bovine, Guinea Pig, Hamster, Human, Monkey, Mouse, Rabbit, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR