Imprimer

Anti-HSPA5 Rabbit Polyclonal Antibody [clone: N/A]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326416EA 593 EUR
ORIGTA326416
Anti-HSPA5 Rabbit Polyclonal Antibody [clone: N/A]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: HSPA5
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: canine, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR