Imprimer

Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326419EA 584 EUR
ORIGTA326419
Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]
Anticorps
Recommended Dilutions: 0.5?g/ml

Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: 9G10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Bovine, canine, Chicken, Guinea Pig, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR