Imprimer

Anti-TP53 Rabbit Polyclonal Antibody [clone: Poly6142]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327919EA 512 EUR
ORIGTA327919
Anti-TP53 Rabbit Polyclonal Antibody [clone: Poly6142]
Anticorps
Type: Primary
Antigen: TP53
Clonality: Polyclonal
Clone: Poly6142
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR