Imprimer

Anti-BCL2 Mouse Monoclonal Antibody [clone: BCL/10C4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327922EA 479 EUR
ORIGTA327922
Anti-BCL2 Mouse Monoclonal Antibody [clone: BCL/10C4]
Anticorps
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: BCL/10C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR