Imprimer

Anti-IRF3 Mouse Monoclonal Antibody [clone: SL-12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327933EA 466 EUR
ORIGTA327933
Anti-IRF3 Mouse Monoclonal Antibody [clone: SL-12]
Type: Primary
Antigen: IRF3
Clonality: Monoclonal
Clone: SL-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR