Imprimer

Anti-PARP1 Mouse Monoclonal Antibody [clone: 5A5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327935EA 479 EUR
ORIGTA327935
Anti-PARP1 Mouse Monoclonal Antibody [clone: 5A5]
Anticorps
Type: Primary
Antigen: PARP1
Clonality: Monoclonal
Clone: 5A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR