Imprimer

Anti-STRN4 Mouse Monoclonal Antibody [clone: S88-64]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326434EA 0 EUR
ORIGTA326434
Anti-STRN4 Mouse Monoclonal Antibody [clone: S88-64]
Anticorps
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: STRN4
Clonality: Monoclonal
Clone: S88-64
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR