Imprimer

Anti-DNMT1 Mouse Monoclonal Antibody [clone: 60B1220.1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326439EA 692 EUR
ORIGTA326439
Anti-DNMT1 Mouse Monoclonal Antibody [clone: 60B1220.1]
Anticorps
Recommended Dilutions: Western Blot: 2-4?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: DNMT1
Clonality: Monoclonal
Clone: 60B1220.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, zebraFish
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR