Imprimer

Anti-DNMT1 Mouse Monoclonal Antibody [clone: 4G11-C7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326440EA 645 EUR
ORIGTA326440
Anti-DNMT1 Mouse Monoclonal Antibody [clone: 4G11-C7]
Anticorps
Recommended Dilutions: Western Blot: 2-4?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: DNMT1
Clonality: Monoclonal
Clone: 4G11-C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, zebraFish
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR