Imprimer

Anti-ANK2 Mouse Monoclonal Antibody [clone: S105-17]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326444EA 574 EUR
ORIGTA326444
Anti-ANK2 Mouse Monoclonal Antibody [clone: S105-17]
Recommended Dilutions: Western Blot: 1?g/ml

Type: Primary
Antigen: ANK2
Clonality: Monoclonal
Clone: S105-17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR