Imprimer

Anti-UBB Mouse Monoclonal Antibody [clone: 5B9-B3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326455EA 604 EUR
ORIGTA326455
Anti-UBB Mouse Monoclonal Antibody [clone: 5B9-B3]
Anticorps
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: UBB
Clonality: Monoclonal
Clone: 5B9-B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Bovine, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR