Imprimer

Anti-AHSA1 Rat Monoclonal Antibody [clone: 25F2.D9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326464EA 665 EUR
ORIGTA326464
Anti-AHSA1 Rat Monoclonal Antibody [clone: 25F2.D9]
Anticorps
Type: Primary
Antigen: AHSA1
Clonality: Monoclonal
Clone: 25F2.D9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR