Imprimer

Anti-F2R Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA590107EA 638 EUR
ORIGTA590107
Anti-F2R Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:5000-1:20000; ELISA: 1:100-1:2000

Type: Primary
Antigen: F2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR