Imprimer

Anti-LCP1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA590120EA 568 EUR
ORIGTA590120
Anti-LCP1 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot 1:5000-20000, IHC 1:150, ELISA 1:100-1:2000

Type: Primary
Antigen: LCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Dog, Rat, Monkey, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR