Imprimer

Anti-NFKB1 Mouse Monoclonal Antibody [clone: 4D1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327985EA 479 EUR
ORIGTA327985
Anti-NFKB1 Mouse Monoclonal Antibody [clone: 4D1]
Anticorps
Type: Primary
Antigen: NFKB1
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR