Imprimer

Anti-CDKN1B Rabbit Polyclonal Antibody [clone: Poly6029]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327987EA 466 EUR
ORIGTA327987
Anti-CDKN1B Rabbit Polyclonal Antibody [clone: Poly6029]
Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone: Poly6029
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR