Imprimer

Anti-MCM2 Rabbit Polyclonal Antibody [clone: Poly6022]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327997EA 182 EUR
ORIGTA327997
Anti-MCM2 Rabbit Polyclonal Antibody [clone: Poly6022]
Type: Primary
Antigen: MCM2
Clonality: Polyclonal
Clone: Poly6022
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR