Imprimer

Anti-AKT1 Rabbit Polyclonal Antibody [clone: Poly6490]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA328006EA 874 EUR
ORIGTA328006
Anti-AKT1 Rabbit Polyclonal Antibody [clone: Poly6490]
Anticorps
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone: Poly6490
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR