Imprimer
Anti-STAT2 Rabbit Polyclonal Antibody [clone: Poly6245]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA328010EA 564 EUR
ORIGTA328010
Anti-STAT2 Rabbit Polyclonal Antibody [clone: Poly6245]
Anticorps
Type: Primary
Antigen: STAT2
Clonality: Polyclonal
Clone: Poly6245
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human