Imprimer

Anti-TCEB1 Rabbit Polyclonal Antibody [clone: Poly6131]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA328011EA 525 EUR
ORIGTA328011
Anti-TCEB1 Rabbit Polyclonal Antibody [clone: Poly6131]
Anticorps
Type: Primary
Antigen: TCEB1
Clonality: Polyclonal
Clone: Poly6131
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR