Imprimer

Anti-GABRB3 Mouse Monoclonal Antibody [clone: S87-25]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326474EA 692 EUR
ORIGTA326474
Anti-GABRB3 Mouse Monoclonal Antibody [clone: S87-25]
Anticorps
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: GABRB3
Clonality: Monoclonal
Clone: S87-25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR