Imprimer

Anti-HSF2 Rat Monoclonal Antibody [clone: 3E2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326477EA 519 EUR
ORIGTA326477
Anti-HSF2 Rat Monoclonal Antibody [clone: 3E2]
Anticorps
Recommended Dilutions: 4?g/ml

Type: Primary
Antigen: HSF2
Clonality: Monoclonal
Clone: 3E2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Bovine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit. Dog, Rat, Sheep
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR