Imprimer

Anti-KCNMB4 Mouse Monoclonal Antibody [clone: S18A-3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326489EA 645 EUR
ORIGTA326489
Anti-KCNMB4 Mouse Monoclonal Antibody [clone: S18A-3]
Anticorps
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: KCNMB4
Clonality: Monoclonal
Clone: S18A-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat.
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR