Imprimer

Anti-MDC1 Mouse Monoclonal Antibody [clone: P2B11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326493EA 582 EUR
ORIGTA326493
Anti-MDC1 Mouse Monoclonal Antibody [clone: P2B11]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MDC1
Clonality: Monoclonal
Clone: P2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chimpanzee, Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR