Imprimer

Anti-PCLO Mouse Monoclonal Antibody [clone: 6H9-B6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326497EA 630 EUR
ORIGTA326497
Anti-PCLO Mouse Monoclonal Antibody [clone: 6H9-B6]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PCLO
Clonality: Monoclonal
Clone: 6H9-B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR