Imprimer

Anti-Syt10 Mouse Monoclonal Antibody [clone: S269-73]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326502EA 574 EUR
ORIGTA326502
Anti-Syt10 Mouse Monoclonal Antibody [clone: S269-73]
Type: Primary
Antigen: Syt10
Clonality: Monoclonal
Clone: S269-73
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR