Imprimer

Anti-Dlg4 Mouse Monoclonal Antibody [clone: 6G6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326527EA 603 EUR
ORIGTA326527
Anti-Dlg4 Mouse Monoclonal Antibody [clone: 6G6]
Anticorps
Recommended Dilutions: Western Blot: 1:250

Type: Primary
Antigen: Dlg4
Clonality: Monoclonal
Clone: 6G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR