Imprimer

Anti-Gabbr2 Mouse Monoclonal Antibody [clone: S81-2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326531EA 623 EUR
ORIGTA326531
Anti-Gabbr2 Mouse Monoclonal Antibody [clone: S81-2]
Anticorps
Type: Primary
Antigen: Gabbr2
Clonality: Monoclonal
Clone: S81-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR