Imprimer

Anti-Hspa5 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326546EA 593 EUR
ORIGTA326546
Anti-Hspa5 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Hspa5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR