Imprimer

Anti-MYD88 Rabbit Polyclonal Antibody [clone: Poly6489]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327950EA 466 EUR
ORIGTA327950
Anti-MYD88 Rabbit Polyclonal Antibody [clone: Poly6489]
Type: Primary
Antigen: MYD88
Clonality: Polyclonal
Clone: Poly6489
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR