Imprimer

Anti-CCL18 Mouse Monoclonal Antibody [clone: PO69]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327959EA 466 EUR
ORIGTA327959
Anti-CCL18 Mouse Monoclonal Antibody [clone: PO69]
Type: Primary
Antigen: CCL18
Clonality: Monoclonal
Clone: PO69
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR