Imprimer

Anti-SOX2 Rabbit Polyclonal Antibody [clone: Poly6308]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327960EA 661 EUR
ORIGTA327960
Anti-SOX2 Rabbit Polyclonal Antibody [clone: Poly6308]
Anticorps
Type: Primary
Antigen: SOX2
Clonality: Polyclonal
Clone: Poly6308
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR