Imprimer

Anti-HIST3H3 Rat Monoclonal Antibody [clone: HTA28]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327973EA 874 EUR
ORIGTA327973
Anti-HIST3H3 Rat Monoclonal Antibody [clone: HTA28]
Anticorps
Type: Primary
Antigen: HIST3H3
Clonality: Monoclonal
Clone: HTA28
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR