Imprimer

Anti-Kcnd2 Mouse Monoclonal Antibody [clone: S57-1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326559EA 645 EUR
ORIGTA326559
Anti-Kcnd2 Mouse Monoclonal Antibody [clone: S57-1]
Anticorps
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: Kcnd2
Clonality: Monoclonal
Clone: S57-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR