Imprimer

Anti-Kcnip1 Mouse Monoclonal Antibody [clone: S55-7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326561EA 623 EUR
ORIGTA326561
Anti-Kcnip1 Mouse Monoclonal Antibody [clone: S55-7]
Anticorps
Recommended Dilutions: Immunochemistry: 0.1-1?g/ml

Type: Primary
Antigen: Kcnip1
Clonality: Monoclonal
Clone: S55-7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR