Imprimer

Anti-Kcnip2 Mouse Monoclonal Antibody [clone: S60-73]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326563EA 623 EUR
ORIGTA326563
Anti-Kcnip2 Mouse Monoclonal Antibody [clone: S60-73]
Anticorps
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: Kcnip2
Clonality: Monoclonal
Clone: S60-73
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR