Imprimer

Anti-Kcnt1 Mouse Monoclonal Antibody [clone: S3-26]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326566EA 645 EUR
ORIGTA326566
Anti-Kcnt1 Mouse Monoclonal Antibody [clone: S3-26]
Anticorps
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: Kcnt1
Clonality: Monoclonal
Clone: S3-26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR