Imprimer

Anti-Lamp1 Mouse Monoclonal Antibody [clone: Ly1C6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326567EA 671 EUR
ORIGTA326567
Anti-Lamp1 Mouse Monoclonal Antibody [clone: Ly1C6]
Anticorps
Recommended Dilutions: 1?g/ml

Type: Primary
Antigen: Lamp1
Clonality: Monoclonal
Clone: Ly1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR