Imprimer

Anti-Mmp9 Mouse Monoclonal Antibody [clone: S51-82]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326571EA 645 EUR
ORIGTA326571
Anti-Mmp9 Mouse Monoclonal Antibody [clone: S51-82]
Anticorps
Type: Primary
Antigen: Mmp9
Clonality: Monoclonal
Clone: S51-82
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR