Imprimer

Anti-Panx2 Mouse Monoclonal Antibody [clone: S121A-31]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326573EA 574 EUR
ORIGTA326573
Anti-Panx2 Mouse Monoclonal Antibody [clone: S121A-31]
Recommended Dilutions: Western Blot: 1?g/ml

Type: Primary
Antigen: Panx2
Clonality: Monoclonal
Clone: S121A-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR