Imprimer

Anti-DNMT3B Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327439EA 524 EUR
ORIGTA327439
Anti-DNMT3B Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: DNMT3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR