Imprimer

Anti-PDCD6IP Mouse Monoclonal Antibody [clone: 3A9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA328026EA 500 EUR
ORIGTA328026
Anti-PDCD6IP Mouse Monoclonal Antibody [clone: 3A9]
Anticorps
Type: Primary
Antigen: PDCD6IP
Clonality: Monoclonal
Clone: 3A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: frog, Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR