Imprimer

Anti-PAX5 Rat Monoclonal Antibody [clone: 1H9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA328030EA 525 EUR
ORIGTA328030
Anti-PAX5 Rat Monoclonal Antibody [clone: 1H9]
Anticorps
Type: Primary
Antigen: PAX5
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR